missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Latanoprost, TCI America™

$400.28 - $1,396.42

Chemical Identifiers

CAS 130209-82-4
Molecular Formula C26H40O5
Molecular Weight (g/mol) 432.601
MDL Number MFCD00216074
InChI Key GGXICVAJURFBLW-WRBHGYNOSA-N
Synonym isopropyl 7-1r,2r,3r,5s-3,5-dihydroxy-2-r-3-hydroxy-5-phenylpentyl cyclopentyl hept-5-enoate
PubChem CID 86769173
IUPAC Name propan-2-yl (E)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-(3-hydroxy-5-phenylpentyl)cyclopentyl]hept-5-enoate
SMILES CC(C)OC(=O)CCCC=CCC1C(CC(C1CCC(CCC2=CC=CC=C2)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L026210MG View Documents TCI America
L026210MG
10mg Each for $400.28
 
L026250MG View Documents TCI America
L026250MG
50mg Each for $1,396.42
 
Specifications

Chemical Identifiers

130209-82-4
432.601
GGXICVAJURFBLW-WRBHGYNOSA-N
86769173
CC(C)OC(=O)CCCC=CCC1C(CC(C1CCC(CCC2=CC=CC=C2)O)O)O
C26H40O5
MFCD00216074
isopropyl 7-1r,2r,3r,5s-3,5-dihydroxy-2-r-3-hydroxy-5-phenylpentyl cyclopentyl hept-5-enoate
propan-2-yl (E)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-(3-hydroxy-5-phenylpentyl)cyclopentyl]hept-5-enoate

Specifications

Yellow
130209-82-4
MFCD00216074
GGXICVAJURFBLW-WRBHGYNOSA-N
propan-2-yl (E)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-(3-hydroxy-5-phenylpentyl)cyclopentyl]hept-5-enoate
86769173
Clear Liquid at 20°C
Latanoprost
10mg
C26H40O5
isopropyl 7-1r,2r,3r,5s-3,5-dihydroxy-2-r-3-hydroxy-5-phenylpentyl cyclopentyl hept-5-enoate
CC(C)OC(=O)CCCC=CCC1C(CC(C1CCC(CCC2=CC=CC=C2)O)O)O
432.601
432.60
≥98.0% (HPLC)
Safety and Handling

Safety and Handling

RTECSNumber : MJ9669550

TSCA : No

Recommended Storage : Freezer