Please login to your online account to display your discounted pricing

LAT Mouse anti-Human, Clone: LAT.10-17 (10-17), eBioscience™

Mouse Monoclonal Antibody

$143.55 - $303.05

Specifications

Antigen LAT
Clone LAT.10-17 (10-17)
Host Species Murine
Gene Alias Linker for activation of T-cells
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122896

 
life technologies
LS14996780
25μg Each for $143.55

5013936

 
life technologies
LS14996782
100μg Each for $303.05
Description & Specifications

Specifications

Antigen LAT
Clone LAT.10-17 (10-17)
Host Species Murine
Gene Alias Linker for activation of T-cells
Species Reactivity Human
Applications Immunoprecipitation
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

This LAT.10-17 (10-17) monoclonal antibody reacts with human Linker for Activation of T cells (LAT), which is essential for T cell receptor (TCR)-mediated signal transduction. Characterized as an integral transmembrane protein, this ∽37kDa molecule does not possess a true extracellular domain. Upon TCR/CD3 engagement, LAT localizes to lipid rafts and is tyrosine phosphorylated by ZAP-70, leading to the recruitment and activation of other downstream signaling molecules such as Grb-2, PLCγ, and PI3K. This adapter molecule has been shown to be required for T cell activation, as well as thymocyte and T regulatory cell development. In humans, LAT is expressed on peripheral blood lymphocytes, lymph nodes, and tonsil. Studies in mice have shown that LAT is expressed in thymocytes, T cells, mast cells, natural killer cells, megakaryocytes, platelets, and early B cells. This antibody does not crossreact with the mouse antigen.