LASER Blox™ Laser Pointer Multi 3 Pack
Demonstrate and study reflection, refraction, diffraction, and advanced geometric optics.
Manufacturer:  Fisher Science Education RGVLB05
Catalog No. S12843
Description
Description
  • With red 635nm, green 532nn and violet 405nm LASER Bloxtrademark (TM)
  • Stack them to create ray box, stick them to white board with included mount/use them as separate units
  • Generate visible line with included line generating lens or use as point source
  • Includes: Durable snap foam case, AAA Batteries, line generating lens
Specifications
Specifications
LASER Blox Laser Pointer Multi 3 Pack