missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dynalon™ Large Polyethylene Wide-Mouth Bottles

Available in two useful sizes.

Manufacturer:  Dynalon™ DAL600335

 View more versions of this product

Catalog No. S31944B




Includes: White metal screw caps

Description

Description

  • Large, polyethylene utility jars have hundreds of uses around the lab
  • Natural color

Specifications

Specifications

135.2 oz., 4000 mL
135.2 oz.
Natural
White
152 mm
110
4000 mL
Tinfoil
Polyethylene
110 mm
254 mm
Screw
Documents