missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

β-Lapachone, Tocris

DNA topoisomerase I inhibitor

$138.08 - $581.48

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C15H14O3
Formula Weight 242.27
Synonym 3,4-Dihydro-2,2-dimethyl-2H-naphtho[1,2-b]pyran-5,6-dione
Chemical Name or Material β-Lapachone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
129310 R & D Systems
RDS129310
10mg Each for $138.08
129350 R & D Systems
RDS129350
50mg Each for $581.48
Specifications Description & Specifications

Chemical Identifiers

>98%
242.27
β-Lapachone
C15H14O3
3,4-Dihydro-2,2-dimethyl-2H-naphtho[1,2-b]pyran-5,6-dione

Specifications

4707-32-8
C15H14O3
3,4-Dihydro-2,2-dimethyl-2H-naphtho[1,2-b]pyran-5,6-dione
β-Lapachone
>98%
242.27
10mg

Safety and Handling

Recommended Storage : Store at +4°C