Please login to your online account to display your discounted pricing

β-Lapachone, Tocris


DNA topoisomerase I inhibitor

Quantity 50mg

Manufacturer: r & d systems  RDS129350

 View more versions of this product

Catalog No. 129350

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 242.27
Quantity 50mg
Assay Percent Range >98%
Molecular Formula C15H14O3
CAS 4707-32-8
Chemical Name or Material β-Lapachone
Synonym 3,4-Dihydro-2,2-dimethyl-2H-naphtho[1,2-b]pyran-5,6-dione
Recommended Storage Store at +4°C

Specifications

Chemical Name or Material β-Lapachone
Assay Percent Range >98%
Molecular Formula C15H14O3
Formula Weight 242.27
Synonym 3,4-Dihydro-2,2-dimethyl-2H-naphtho[1,2-b]pyran-5,6-dione
Accessories

Store at +4°C