LanthaScreen™ Tb-PY20 Antibody DFS Item
LanthaScreen™ Tb-PY20 Antibody
Manufacturer:  Life Technologies LSPV3529
 View more versions of this product
Catalog No. PV3529
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
A general reagent to monitor tyrosine kinase activity, the pY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate.
For use in combination with Fluorescein-poly-GT (PV3610) or Fluorescein-poly-GAT (PV3611) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
Specifications
Tyrosine kinases
0.54mg/mL
150kDa
RUO, For research use only
Primary
1 Each
Biochemical Kinase Assays, Drug Discovery and Development, Kinase Biology, Pharma and Biopharma, Target and Lead Identification and Validation
Tb (Terbium), lanthanide chelates
1mg
–20°C
Primary
Documents
For Research Use Only. Not for use in diagnostic procedures.