missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
LanthaScreen™ Tb-pERM (pLRRKtide) Antibody
LanthaScreen™ Tb-pERM (pLRRKtide) Antibody
$7620.00 - $40600.00
Specifications
Antigen | Serine/threonine kinase |
---|---|
Concentration | 0.52mg/mL |
Molecular Weight of Antigen | 150kDa |
Packaging | 1 Each |
Primary or Secondary | Primary |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PV4897
|
Thermo Fisher Scientific
PV4897 |
25 μg |
Each for $7,620.00
|
|
|||||
PV4898
|
Thermo Fisher Scientific
PV4898 |
1 mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
A phospho specific antibody directed to the ezrin⁄radixin⁄moesin (ERM) peptide that is a substrate for LRRK2 (Leucine-rich repeat kinase 2) and LRRK2 mutants. Mutations in LRRK2 have been found to be related to late-onset Parkinson's disease.For use in combination with the Fluorescein-ERM (LRRKtide) substrate (PV4901) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
Serine/threonine kinase | |
150kDa | |
Primary | |
Kinase Assay | |
Liquid |
0.52mg/mL | |
1 Each | |
Terbium-labeled pERM (pLRRKtide) antibody in HEPES buffered saline. Store at -20°C. | |
Tb (Terbium)/Lanthanide Chelates | |
RUO, RUO |
For Research Use Only. Not for use in diagnostic procedures.