LanthaScreen™ Tb-pATF2 (pThr71) Antibody DFS Item
LanthaScreen™ Tb-pATF2 (pThr71) Antibody
Manufacturer:  Life Technologies LSPV4448
 View more versions of this product
Catalog No. PV4448
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
A member of the leucine zipper family of DNA binding proteins, ATF2 (activating transcription factor 2) is phosphorylated at Thr71 by members of the JNK and p38 families of kinases.
For use with the GFP-ATF2 physiological protein substrate (PV4445) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
Specifications
Biochemical Kinase Assays, Drug Discovery and Development, Kinase Biology
LanthaScreen™ Tb-pATF2 Antibody, 1mg, Ser/Thr kinases Antigen, Human Species Reactivity, Tb (Terbium) Conjugate, Used For Biochemical Kinase Assays, Drug Discovery and Development, Kinase Biology
1mg
–20°C
Primary
Tb (Terbium)
Liquid
RUO, For research use only
Primary
1 Each
Documents
For Research Use Only. Not for use in diagnostic procedures.