missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

LanthaScreen™ Tb-p4E-BP1 (pThr46) Antibody Kit DFS Item

LanthaScreen™ Tb-p4E-BP1 (pThr46) Antibody Kit

$5,600.00 - $5,600.00

Specifications

Antigen Serine/threonine kinase
Concentration 0.52mg/mL
Molecular Weight of Antigen 150kDa
Packaging 1 Kit(s)
Primary or Secondary Primary
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PV4757
View Documents
Life Technologies
LSPV4757
25μg Each for $5,600.00
 
PV4758
View Documents
Life Technologies
LSPV4758
1mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

A part of the PI3K/AKT Pathway, 4E-BP1 (EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1) is a cytosolic protein that is phosphorylated at Thr46 by mTORC1 upon stimulation of the pathway.

For use in combination with the GFP-4E-BP1 physiological protein substrate (PV4759) to evaluate inhibitors in medium to high throughput screening applications. An optimized mTor assay buffer (PV4794) is also available.
Specifications

Specifications

Serine/threonine kinase
150kDa
Primary
Biochemical Kinase Assays,Drug Discovery and Development,Enzyme and Protein Activity Assays,Kinase Biology,LanthaScreen™ Kinase Activity Assays,Pharma and Biopharma,Protein Assays and Analysis,Protein Biology,Target and Lead Identification and Validation
384-well plate
Primary
0.52mg/mL
1 Kit(s)
–20°C
GFP (EmGFP), Tb (Terbium), lanthanide chelates
For research use only,RUO

For Research Use Only. Not for use in diagnostic procedures.