LanthaScreen™ Tb-p4E-BP1 (pThr46) Antibody Kit DFS Item
LanthaScreen™ Tb-p4E-BP1 (pThr46) Antibody Kit
Manufacturer:  Life Technologies LSPV4758
 View more versions of this product
Catalog No. PV4758
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
A part of the PI3K/AKT Pathway, 4E-BP1 (EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1) is a cytosolic protein that is phosphorylated at Thr46 by mTORC1 upon stimulation of the pathway.
For use in combination with the GFP-4E-BP1 physiological protein substrate (PV4759) to evaluate inhibitors in medium to high throughput screening applications. An optimized mTor assay buffer (PV4794) is also available.
Specifications
Specifications
Serine/threonine kinase
0.52mg/mL
LanthaScreen™ Tb-p4E-BP1 (pThr46) Antibody Kit, 1mg, 0.52mg/mL Concentration, Serine/Threonine Kinases Antigen, Human Species Reactivity, GFP (EmGFP), Tb (Terbium) Conjugate, 340/490nm Excitation, 520nm Emission, 10mM HEPES (pH 7.5), 137mM NaCl and
150kDa
RUO, For research use only
Primary
1 Kit(s)
Biochemical Kinase Assays, Drug Discovery and Development, Enzyme and Protein Activity Assays, Kinase Biology, LanthaScreen™ Kinase Activity Assays, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
GFP (EmGFP), Tb (Terbium), lanthanide chelates
384-well plate
1mg
–20°C
Primary
Documents
For Research Use Only. Not for use in diagnostic procedures.