LanthaScreen™ Tb-IKBa pSer32 Antibody Kit DFS Item
LanthaScreen™ Tb-IKBa pSer32 Antibody Kit
Manufacturer:  Life Technologies LSPV3563
 View more versions of this product
Catalog No. PV3563
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
An essential component of the canonical NF-kB pathway, phosphorylation of IkappaBalpha at Ser32 by IKK preceedes the transactivation of NF-kappaB.
For use in combination with the Fluorescein-IKK peptide substrate (PV3507) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
Specifications
Serine/threonine kinase
FITC (Fluorescein), Tb (Terbium)
150kDa
RUO, For research use only
Primary
1147, 2869, 3551
Biochemical Kinase Assays, Drug Discovery and Development, lanthaScreen™, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation, Validation Kinase Biology Kinase Antibodies and Immunoassays
384-well plate
1mg
–20°C
Primary
1 Kit(s)
Documents
For Research Use Only. Not for use in diagnostic procedures.