LanthaScreen™ Tb-IKBa pSer32 Antibody Kit
LanthaScreen™ Tb-IKBa pSer32 Antibody Kit
Supplier: Thermo Fisher Scientific PV3562
Description
An essential component of the canonical NF-kB pathway, phosphorylation of IkappaBalpha at Ser32 by IKK preceedes the transactivation of NF-kappaB.For use in combination with the Fluorescein-IKK peptide substrate (PV3507) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
I kappa B alpha, phospho (Ser 32) | |
Tb (Terbium)/FITC | |
25 μg | |
Kinase Biology | |
1 Each | |
384-well Plate |
Kinase Assay, Enzyme and Protein Activity Assays, Kinase Assay, Protein Assay | |
LanthaScreen™ Tb-IKBa pSer32 Antibody Kit, 25μg, I kappa B Alpha, Phospho (Ser 32) Antigen, Immunogen, Host, Human Species Reactivity, FITC (Fluorescein), Tb (Terbium) Conjugate Isotype, HEPES Buffered Saline Storage Buffer, Used For Biochemical | |
RUO, RUO | |
Primary | |
–20°C |
For Research Use Only. Not for use in diagnostic procedures.