LanthaScreen™ Tb-anti-phospho-p53 [pSer15] Antibody DFS Item
LanthaScreen™ Tb-anti-phospho-p53 [pSer15] Antibody
Manufacturer:  Life Technologies LSPV5129
 View more versions of this product
Catalog No. PV5129
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
The p53 tumor suppressor protein is an essential component of many different signaling pathways. The LanthaScreen™ antibody detects the phosphorylation of p53 at Ser15 by a range of kinases including DNA-PK, ATM, and ATR.
For use in combination with the Fluorescein-p53 (Ser15) peptide substrate (PV5132) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
Specifications
Ser/Thr kinases
Tb (Terbium)
150kDa
RUO, For research use only
Primary
Monoclonal
Oncology and Genetic Disease Research, Pharma and Biopharma, Target and Lead Identification and Validation
Phosphorylation of p53
1mg
–20°C
Primary
1 Each
Documents
For Research Use Only. Not for use in diagnostic procedures.