missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ LanthaScreen™ Eu-Streptavidin DFS Item

LanthaScreen™ Eu-Streptavidin

$1,310.00 - $1,310.00

Specifications

Molecular Weight 55kDa
Product Name LanthaScreen™ Eu-Streptavidin
Storage Buffer 10mM HEPES (pH 7.5), 137mM NaCl and 2.7mM KCl (HEPES buffered saline)
Storage Requirements –20°C
Concentration 0.25mg/mL
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PV5899 DFS Item
View Documents
Invitrogen™
LSPV5899
25μg
Each for $1,310.00
 
PV6025 DFS Item
View Documents
Invitrogen™
LSPV6025
1mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

A general reagent for use in assay development, the Europium-Streptavidin binds to biotinylated peptide or physiological protein substrates.
Specifications

Specifications

55kDa
10mM HEPES (pH 7.5), 137mM NaCl and 2.7mM KCl (HEPES buffered saline)
0.25mg/mL
LanthaScreen™ Eu-Streptavidin
LanthaScreen™ Eu-Streptavidin
–20°C
Frozen
SDS

For Research Use Only. Not for use in diagnostic procedures.