missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

LanthaScreen™ Eu-anti-His Tag Antibody Kit DFS Item

LanthaScreen™ Eu-anti-His Tag Antibody Kit

Manufacturer:  Life Technologies LSPV5597

 View more versions of this product

Catalog No. PV5597

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

LanthaScreen™ europium-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Eu chelates negatively impacts protein activity. These antibodies can be used with our Kinase Tracers to conduct LanthaScreen™ Eu Kinase Binding Assays.
Specifications

Specifications

Europium (Eu), Lanthanide Chelates
His
340⁄665
High-throughput Compatible
Europium
Dry Ice
TR-FRET
0.25 mg/mL, 0.25 mg/ml, 0.25 mg⁄ml
Liquid
1mg
–20°C
Primary
150 kDa
Europium-labeled anti-His tag antibody in HEPES buffered saline. Store at -20°C.
Microplate Reader
Lanthanide Chelates
LanthaScreen®
His
Biochemical Kinase Assays,Drug Discovery and Development,Kinase Biology,LanthaScreen™ Kinase Activity Assays,Pharma and Biopharma,Target and Lead Identification and Validation
Europium
150kDa
For research use only,RUO
Primary
1 Kit(s)
SDS
Documents

For Research Use Only. Not for use in diagnostic procedures.