missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
LanthaScreen™ Eu-anti-His Tag Antibody Kit
LanthaScreen™ Eu-anti-His Tag Antibody Kit
Supplier: Thermo Fisher Scientific PV5596
Description
LanthaScreen™ europium-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Eu chelates negatively impacts protein activity. These antibodies can be used with our Kinase Tracers to conduct LanthaScreen™ Eu Kinase Binding Assays.Specifications
His | |
0.25 mg/mL, 0.25 mg/ml, 0.25 mg⁄ml | |
150kDa | |
RUO, RUO | |
Primary | |
Europium-labeled 6×His antibody is supplied in HEPES-buffered saline. Store at -20°C. | |
Liquid | |
340⁄665 | |
Dry Ice | |
His | |
LanthaScreen™ |
Kinase Assay, Kinase Assay | |
Europium | |
25 μg | |
Kinase Biology | |
1 Kit(s) | |
150 kDa | |
Biochemical binding/kinetic | |
Microplate Reader | |
CSF1R, PRKCH, PRKCQ, GSK3B, FGFR1, TEC, CAMK2D, RIPK2, FYN, AURKA, NUAK1, FLT3, KDR, CAMK2A, RPS6KA3, AXL, FGR, CAMKK1, CDK9/Cyclin T1, TBK1, PAK6, PDK1, FLT3 D835Y, BTK, FES, PRKACA, PTK6 | |
TR-FRET |
For Research Use Only. Not for use in diagnostic procedures.