missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
LanthaScreen™ Eu-anti-GST Antibody
LanthaScreen™ Eu-anti-GST Antibody
Supplier: Thermo Fisher Scientific PV5595
Description
LanthaScreen™ europium-labeled anti-GST goat antibodies are used to indirectly label proteins tagged with GST. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Eu chelates negatively impacts protein activity. These antibodies can be used with our Kinase Tracers to conduct LanthaScreen™ Eu Kinase Binding Assays.Specifications
GST | |
0.25 mg/mL, 0.25 mg/ml, 0.25 mg⁄ml | |
150kDa | |
RUO, RUO | |
Primary | |
Europium-labeled anti-GST antibody is supplied in HEPES-buffered saline. Store at -20°C. | |
Liquid | |
Microplate Reader | |
GST | |
LanthaScreen™ |
Kinase Assay, Kinase Assay | |
Europium | |
1 mg | |
Pharma and Biopharma | |
1 Kit(s) | |
150 kDa | |
340⁄665 | |
Dry Ice | |
TR-FRET |
For Research Use Only. Not for use in diagnostic procedures.