missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ LanthaScreen™ Amine Reactive Tb Chelate DFS Item

LanthaScreen™ Amine Reactive Tb Chelate

$534.00 - $2,570.00

Specifications

Molecular Weight 1050.88g/mol
Storage Requirements –20°C
Product Type LanthaScreen™ Amine Reactive Tb Chelate
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PV3583
View Documents
Invitrogen™
LSPV3583
10μg Each for $534.00
 
PV3582
View Documents
Invitrogen™
LSPV3582
100μg Each for $2,570.00
 
PV3581
View Documents
Invitrogen™
LSPV3581
1mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

As part of the LanthaScreen™ TR-FRET toolbox of assay reagents, LanthaScreen™ Amine Reactive Tb Chelate labeling reagent is available for assay development. The amine-reactive, isothiocyanate group conjugates to virtually any peptide or protein that contains one or more accessible amine moieties, such as the amine group found on a lysine residue or an unmodified N-terminal amine group of a peptide or protein. Amine-modified DNA oligonucleotides can be labeled as well. Energy transfer from the terbium donor to a suitable acceptor such as fluorescein is readily detected by monitoring an increase in acceptor fluorescence intensity. See the user guide for application-based protocols for labeling peptides containing free amino groups, amine-modified oligonucleotides, and IgG antibodies.
Specifications

Specifications

1050.88g/mol
–20°C
LanthaScreen™ Amine Reactive Tb Chelate

For Research Use Only. Not for use in diagnostic procedures.