Learn More
Lanthanum trifluoromethanesulfonate, anhydrous, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1965714
Description
Lanthanum trifluoromethanesulfonate is used in the Aldol reaction of silyl enol ethers with aldehydes. It is an effective catalyst for the Baylis-Hillman reaction. It also catalyzes Michael additions to methyl vinyl ketone in water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsLanthanum trifluoromethanesulfonate is used in the Aldol reaction of silyl enol ethers with aldehydes. It is an effective catalyst for the Baylis-Hillman reaction. It also catalyzes Michael additions to methyl vinyl ketone in water.
Solubility
Soluble in water.
Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with bases. It is hygroscopic in nature. Store under dry inert gas.
Specifications
Lanthanum trifluoromethanesulfonate | |
52093-26-2 | |
(CF3SO3)3La | |
25 g | |
9009103 | |
lanthanum iii trifluoromethanesulfonate, lanthanum triflate, lanthanum trifluoromethanesulfonate, la otf 3, lanthanum iii triflate, lanthanum tris trifluoromethanesulfonate, lanthanum 3+ tritriflate, lanthanum 3+ ; trifluoromethanesulfonate, trifluoromethanesulfonic acid lanthanum salt, tris trifluoromethanesulfonato lanthanum | |
WGJJZRVGLPOKQT-UHFFFAOYSA-K | |
lanthanum(3+);trifluoromethanesulfonate | |
2733938 | |
99% |
anhydrous | |
C3F9LaO9S3 | |
MFCD00077560 | |
UN3261 | |
Hygroscopic | |
Soluble in water. | |
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[La+3] | |
586.095 | |
586.1 |
Chemical Identifiers
52093-26-2 | |
586.095 | |
WGJJZRVGLPOKQT-UHFFFAOYSA-K | |
2733938 | |
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[La+3] |
C3F9LaO9S3 | |
MFCD00077560 | |
lanthanum iii trifluoromethanesulfonate, lanthanum triflate, lanthanum trifluoromethanesulfonate, la otf 3, lanthanum iii triflate, lanthanum tris trifluoromethanesulfonate, lanthanum 3+ tritriflate, lanthanum 3+ ; trifluoromethanesulfonate, trifluoromethanesulfonic acid lanthanum salt, tris trifluoromethanesulfonato lanthanum | |
lanthanum(3+);trifluoromethanesulfonate |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only