Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum titanium oxide

Chemical Identifiers

Chemical Name or Material Lanthanum titanium oxide
CAS 12031-47-9
Formula Weight 485.57
MDL Number MFCD00016167
Synonym Lanthanum titanate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1821322

  SDS
alfa aesar™
1821322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1821336

  SDS
alfa aesar™
1821336
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 485.57
Chemical Name or Material Lanthanum titanium oxide
TSCA TSCA
Odor Odorless
Physical Form Powder
MDL Number MFCD00016167
CAS 12031-47-9
Synonym Lanthanum titanate
Color White
Quantity 100g