Please login to your online account to display your discounted pricing

Lanthanum Standard, 1000μg/mL in 2-5% HNO3 (Assurance Grade), SPEX CertiPrep™


ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Quantity 125mL
Packaging Triple-leached LDPE bottle

Manufacturer: spex certiprep llc  PLLA22Y

 View more versions of this product

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Catalog No. PLLA2-2Y

  • / Each
 

Description & Specifications

Chemical Identifiers

Concentration or Composition (by Analyte or Components) Matrix&tab;Concentration&tab;Cat. No.
2-5% HNO3&tab;1000μg/mL&tab;PLLA2-2Y, -2X
Quantity 125mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Health Hazard 1 Causes severe skin burns and eye damage
Packaging Triple-leached LDPE bottle
Recommended Storage Ambient
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
DOT Information Hazardous
Solvent or Matrix 2% HNO3
Purity Grade Plasma Grade
Certificate of Analysis Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
CAS Max % 97.9
CAS 7439-91-0
CAS 7697-37-2
CAS Min % 2.0
CAS Max % 2.0
CAS Min % 97.9
Chemical Name or Material Lanthanum Standard
CAS Min % 0.1
CAS Max % 0.1
CAS 7732-18-5

Specifications

Chemical Name or Material Lanthanum Standard
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
La1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

Causes severe skin burns and eye damage

Ambient