missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum sputtering target, 76.2mm dia x 1.59mm thick

Chemical Identifiers

CAS 7439-91-0
MDL Number MFCD00011066
UN Number UN3208
Chemical Name or Material Lanthanum sputtering target, 76.2mm dia. x 1.59mm thick
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA46064KS SDS Alfa Aesar™
46064KS
1 each N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7439-91-0
MFCD00011066
UN3208
Lanthanum sputtering target, 76.2mm dia. x 1.59mm thick

Specifications

1 each
MFCD00011066
Lanthanum sputtering target, 76.2mm dia. x 1.59mm thick
7439-91-0
UN3208

Safety and Handling

GHS H Statement
H228-H261
Flammable solid.
In contact with water releases flammable gas.

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.

TSCA : TSCA