Please login to your online account to display your discounted pricing

Lanthanum Solution, For Metals, LabChem™

$127.38 - $228.68

Chemical Identifiers

Chemical Name or Material Lanthanum Solution
CAS 7732-18-5
CAS Max % 84.3
CAS 10099-58-8
CAS Max % 8.83
CAS 7647-01-0
CAS Max % 6.87
Name Note For Metals
Linear Formula LaCl3
Formula Weight 245.26
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC160501

 
lab chem
160501
500mL Each for $127.38

LC160502

 
lab chem
160502
1L Each for $228.68
Description & Specifications

Specifications

Formula Weight 245.26
Chemical Name or Material Lanthanum Solution
CAS Max % 6.87
Decomposition Information Hydrogen chloride
Color Clear, colorless to slight yellow
Quantity 500mL
Name Note For Metals
Physical Form Liquid
Linear Formula LaCl3
Density 1.1g/mL
CAS 7732-18-5
UN Number UN1789
Health Hazard 1 Danger
CAS Max % 84.3
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Identification Passes Test
CAS 10099-58-8
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Poly Bottle
CAS Max % 8.83
Solubility Soluble in water
CAS 7647-01-0
Specific Gravity 1.1g/mL
Storage Information Room Temperature
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.