missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum Solution, APHA for Metals, Ricca Chemical

$189.82 - $333.88

Chemical Identifiers

CAS 7732-18-5,7647-01-0,1312-81-8
CAS Min % 72.0,21.35,4.27
CAS Max % 75.0,22.22,4.45
Physical Form Liquid
Chemical Name or Material Lanthanum Solution
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
4285-16 SDS Ricca Chemical Company
RI428516
Poly natural 500mL Each for $189.82
4285-32 Ricca Chemical Company
RI428532
Poly natural 1L Each for $333.88
42854 Ricca Chemical Company
RI42854
Poly natural 120mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
R42850004A Ricca Chemical Company
RIR42850004A
Plastic Bottle 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7647-01-0,1312-81-8
75.0,22.22,4.45
Lanthanum Solution
72.0,21.35,4.27
Liquid

Specifications

Liquid
500mL
7732-18-5,1312-81-8,7647-01-0
75.0,4.45,22.22
Lanthanum Solution
Clear colorless
APHA for Metals
72.0,21.35,4.27
Poly natural