Please login to your online account to display your discounted pricing

Lanthanum Solution, APHA for Metals, Ricca Chemical


Quantity 4L
Packaging Plastic Bottle

Manufacturer: ricca chemical company  RIR42850004A

 View more versions of this product

Catalog No. R42850004A

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 4L
Chemical Name or Material Lanthanum Solution
CAS Min % 72.0
CAS Max % 75.0
CAS Max % 22.22
Methods APHA for Metals
CAS 7647-01-0
Packaging Plastic Bottle
CAS 1312-81-8
CAS Max % 4.45
CAS Min % 4.27
CAS Min % 21.35
CAS 7732-18-5

Specifications

Chemical Name or Material Lanthanum Solution
CAS 7732-18-5
CAS Min % 72.0
CAS Max % 75.0
CAS 7647-01-0
CAS Min % 21.35
CAS Max % 22.22
CAS 1312-81-8
CAS Min % 4.27
CAS Max % 4.45
Physical Form Liquid