missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum Powder, -40 mesh, 99.9% (REO), Alfa Aesar™

Air and moisture

Manufacturer:  Alfa Aesar 4418206

 View more versions of this product

Catalog No. AA4418206



Specifications

Specifications

Lanthanum
7439-91-0
99.9% (REO)
MFCD00011066
FZLIPJUXYLNCLC-UHFFFAOYSA-N
[La]
138.905
CHEBI:33336
Gray to Silver
5g
Gray to silver powder
La
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
Odorless
lanthanum
23926
Powder

Chemical Identifiers

7439-91-0
138.905
FZLIPJUXYLNCLC-UHFFFAOYSA-N
23926
lanthanum
La
MFCD00011066
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
CHEBI:33336
[La]
Safety and Handling

Safety and Handling

H228-H260: Flammable solid. In contact with water releases flammable gases which may ignite spontaneously.

DOTInformation : DOT Class: 4.1, Packaging Group: II

EINECSNumber : 231-099-0

SDS Documents