missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum plasma standard solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13870AD

 View more versions of this product

Catalog No. AA13870AD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Lanthanum
Clear liquid
MFCD00011066
Liquid
Colorless
1312-81-8,7697-37-2,7732-18-5
La2O3 in 5% HNO3
Nitric acid: 5.0%; Lanthanum(III) oxide: 0.12%; Water: 94.88%
23 hPa (17mm Hg) at 20°C
50mL

Chemical Identifiers

1312-81-8
La2O3 in 5% HNO3
MFCD00011066
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

Documents