missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum Oxide, Reagent Grade, LabChem™

$68.69 - $68.69

Chemical Identifiers

CAS 1312-81-8
CAS Max % 100
Linear Formula La2O3
Formula Weight 325.81
Chemical Name or Material Lanthanum Oxide
Grade Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC160409 Lab Chem
LC160409
100g Each for $68.69
Specifications Description & Specifications

Chemical Identifiers

1312-81-8
La2O3
Lanthanum Oxide
100
325.81
Reagent Grade

Specifications

1312-81-8
Powder
325.81
Passes Test
Soluble in water
Room Temperature
4,200°C
100g
Reagent Grade
100
La2O3
6.51g/cm3
Poly Bottle
6.51g/cm3
Fume; Carbon monoxide; Carbon dioxide
White
Lanthanum Oxide

Safety and Handling

GHS H Statement
Causes eye irritation.

GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning