Please login to your online account to display your discounted pricing

Lanthanum Nitrate, 6-Hydrate, BAKER™, J.T.Baker™


A grade of chemicals for laboratory use

Quantity 100g
Packaging Wide Mouth Amber Glass Bottle

Manufacturer: avantor performance  P35405

 View more versions of this product

Catalog No. 02003313

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 433.03
Chemical Name or Material Lanthanum Nitrate
Name Note 6-Hydrate
Molecular Formula La(NO3)3·6H2O
CAS 10277-43-7
Quantity 100g
Packaging Wide Mouth Amber Glass Bottle

Specifications

Chemical Name or Material Lanthanum Nitrate
CAS 10277-43-7
Name Note 6-Hydrate
Molecular Formula La(NO3)3·6H2O
Formula Weight 433.03