missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum Nitrate, 6-Hydrate, BAKER™, J.T.Baker™

A grade of chemicals for laboratory use

$312.07 - $767.36

Chemical Identifiers

CAS 10277-43-7
Molecular Formula La(NO3)3·6H2O
Formula Weight 433.03
Chemical Name or Material Lanthanum Nitrate, 6-Hydrate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
02003313 Avantor Performance
P35405
Wide Mouth Amber Glass Bottle 100g Each for $312.07
02003314 Avantor Performance
P35407
Wide Mouth Amber Glass Bottle 500g Each for $767.36
Specifications Description & Specifications

Chemical Identifiers

10277-43-7
La(NO3)3·6H2O
433.03
Lanthanum Nitrate, 6-Hydrate

Specifications

433.03
La(NO3)3·6H2O
Wide Mouth Amber Glass Bottle
10277-43-7
100g
Lanthanum Nitrate, 6-Hydrate