missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum nickel, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01127118

 View more versions of this product

Catalog No. AA1127118



Description

Description

Lanthanum nickel act as a superconducting intermetallic phase in lanthanum nickel boro-nitride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Specifications

Lanthanum nickel
(REO)
MFCD00198177
50 g
Reacts with water at 0°C.
[Ni].[Ni].[Ni].[Ni].[Ni].[La]
432.37
99.9%
Odorless
Pieces
12196-72-4
LaNi5
UN3178
lanthanum nickel, lani5, lanthanum pentanickel, lanthanum, compd. with nickel 1:1, lanthanum-nickel alloy, acmc-1brhe, lanthanum nickel, reacton, lanthanum nickel powder,-100 mesh, lanthanum nickel eutectic pieces, reacton, hy-stor™ 205 lanthanum-nickel alloy
BUCVQDCWULHLJZ-UHFFFAOYSA-N
lanthanum;nickel
14367487
Reacton™
Air and moisture sensitive

Chemical Identifiers

12196-72-4
432.37
BUCVQDCWULHLJZ-UHFFFAOYSA-N
14367487
[Ni].[Ni].[Ni].[Ni].[Ni].[La]
LaNi5
MFCD00198177
lanthanum nickel, lani5, lanthanum pentanickel, lanthanum, compd. with nickel 1:1, lanthanum-nickel alloy, acmc-1brhe, lanthanum nickel, reacton, lanthanum nickel powder,-100 mesh, lanthanum nickel eutectic pieces, reacton, hy-stor™ 205 lanthanum-nickel alloy
lanthanum;nickel
Safety and Handling

Safety and Handling

GHS H Statement
H372-H351-H228-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Flammable solid.
May cause an allergic skin reaction.

P201-P202-P210-P240-P241-P260-P264b-P270-P281-P285-P304+P341-P308+P313-P342+P311-P370+P378q-P501c

H228-H334-H350-H372

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

EINECSNumber : 235-372-5

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only