missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum nickel, REacton™, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1127190

 View more versions of this product

Catalog No. AA1127190



Description

Description

  • Packaged under argon
Specifications

Specifications

Lanthanum nickel
12196-72-4
LaNi
UN3178
Reacts with water at 0°C.
[Ni].[La]
Odorless
14367487
99.9%
Air and moisture sensitive
5 x 50g
(REO)
MFCD00198177
acmc-1brhe, hy-stor™ 205 lanthanum-nickel alloy, lani5, lanthanum nickel, lanthanum nickel eutectic pieces, reacton, lanthanum nickel powder,-100 mesh, lanthanum nickel, reacton, lanthanum pentanickel, lanthanum, compd. with nickel 1:1, lanthanum-nickel alloy
DOARWPHSJVUWFT-UHFFFAOYSA-N
lanthanum;nickel
197.599
Pieces,Intermetallic,Packaged under argon
Reacton™

Chemical Identifiers

12196-72-4
197.599
DOARWPHSJVUWFT-UHFFFAOYSA-N
14367487
[Ni].[La]
LaNi
MFCD00198177
acmc-1brhe, hy-stor™ 205 lanthanum-nickel alloy, lani5, lanthanum nickel, lanthanum nickel eutectic pieces, reacton, lanthanum nickel powder,-100 mesh, lanthanum nickel, reacton, lanthanum pentanickel, lanthanum, compd. with nickel 1:1, lanthanum-nickel alloy
lanthanum;nickel
Safety and Handling

Safety and Handling

GHS H Statement
H372-H351-H228-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Flammable solid.
May cause an allergic skin reaction.

P201-P202-P210-P240-P241-P260-P264b-P270-P281-P285-P304+P341-P308+P313-P342+P311-P370+P378q-P501c

H228-H334-H350-H372

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

EINECSNumber : 235-372-5

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

Documents