Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum Nickel eutectic pieces, REacton™, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Lanthanum Nickel eutectic pieces
Purity Grade Reacton™
MDL Number MFCD00198177
UN Number UN3178
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1339918

  SDS
alfa aesar™
1339918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Sensitivity Air & Moisture Sensitive
Assay 99.9%
Chemical Name or Material Lanthanum Nickel eutectic pieces
Purity Grade Reacton™
MDL Number MFCD00198177
Physical Form Pieces
Health Hazard 2 GHS H Statement
H350-H228-H317
May cause cancer.
Flammable solid.
May cause an allergic skin reaction.
Quantity 50g
Assay Percent Notes (REO)
Health Hazard 3 GHS P Statement
P210-P261-P280-P363-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 480°C
Avg. Mol. Wt. or Mol. Wt. Range La:Ni; 88:12 wt%
Health Hazard 1 Danger
UN Number UN3178
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P363-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H228-H317
May cause cancer.
Flammable solid.
May cause an allergic skin reaction.