missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum(III) Trifluoromethanesulfonate, TCI America™

Manufacturer:  TCI America T129325G

 View more versions of this product

Catalog No. T129325G



Specifications

Specifications

Lanthanum(III) Trifluoromethanesulfonate
25g
C3F9LaO9S3
1759
WGJJZRVGLPOKQT-UHFFFAOYSA-K
lanthanum(3+);trifluoromethanesulfonate
2733938
Crystal-Powder at 20°C
White
52093-26-2
MFCD00077560
lanthanum iii trifluoromethanesulfonate, lanthanum triflate, lanthanum trifluoromethanesulfonate, la otf 3, lanthanum iii triflate, lanthanum tris trifluoromethanesulfonate, lanthanum 3+ tritriflate, lanthanum 3+ ; trifluoromethanesulfonate, trifluoromethanesulfonic acid lanthanum salt, tris trifluoromethanesulfonato lanthanum
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[La+3]
586.095
586.10
≥98.0% (T)

Chemical Identifiers

52093-26-2
586.095
WGJJZRVGLPOKQT-UHFFFAOYSA-K
2733938
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[La+3]
C3F9LaO9S3
MFCD00077560
lanthanum iii trifluoromethanesulfonate, lanthanum triflate, lanthanum trifluoromethanesulfonate, la otf 3, lanthanum iii triflate, lanthanum tris trifluoromethanesulfonate, lanthanum 3+ tritriflate, lanthanum 3+ ; trifluoromethanesulfonate, trifluoromethanesulfonic acid lanthanum salt, tris trifluoromethanesulfonato lanthanum
lanthanum(3+);trifluoromethanesulfonate
Safety and Handling

Safety and Handling

TSCA : No

Documents