Please login to your online account to display your discounted pricing

Lanthanum(III) trifluoromethanesulfonate, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  34391-0010

 View more versions of this product

Catalog No. AC343910010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 586.1
Quantity 1g
Color White to light beige
Density 1.7g/mL
Chemical Name or Material Lanthanum(III) trifluoromethanesulfonate
Assay Percent Range 99%
Molecular Formula C3F9LaO9S3
MDL Number MFCD00077560
Synonym Trifluoromethanesulfonic acid lanthanum(III) salt
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
CAS 52093-26-2
Elemental Analysis 98.5% min. (ex F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

Specifications

Chemical Name or Material Lanthanum(III) trifluoromethanesulfonate
Assay Percent Range 99%
Molecular Formula C3F9LaO9S3
Formula Weight 586.1
MDL Number MFCD00077560
Synonym Trifluoromethanesulfonic acid lanthanum(III) salt
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents