Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum(III) sulfate hydrate, REacton™, 99.99% (REO)

Chemical Identifiers

Chemical Name or Material Lanthanum(III) sulfate hydrate
CAS 57804-25-8
Purity Grade Reacton™
Formula Weight 566.00 (Anhydrous)
MDL Number MFCD00011070
Merck Index 14,5363
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1319518

  SDS
alfa aesar™
1319518
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1319530

  SDS
alfa aesar™
1319530
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 566.00 (Anhydrous)
TSCA TSCA
Merck Index 14,5363
Purity Grade Reacton™
Sensitivity Hygroscopic
Assay Percent Notes (REO)
CAS 57804-25-8
Quantity 50g
Physical Form Crystalline
Chemical Name or Material Lanthanum(III) sulfate hydrate
Density 2.821g/mL
MDL Number MFCD00011070
Assay 99.99%