missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum(III) sulfate, anhydrous, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 8976018

 View more versions of this product

Catalog No. AA8976018



Specifications

Specifications

Lanthanum (III) sulfate
10099-60-2
(REO)
MFCD00011070
VQEHIYWBGOJJDM-UHFFFAOYSA-H
lanthanum(3+);trisulfate
159259
Powder
Odorless
White
50g
Anhydrous
99.9%
La2O12S3
14,5363
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[La+3].[La+3]
565.979
566
3.60g/mL
Hygroscopic
1150°C (decomposition)

Chemical Identifiers

10099-60-2
565.979
VQEHIYWBGOJJDM-UHFFFAOYSA-H
lanthanum(3+);trisulfate
La2O12S3
MFCD00011070
159259
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[La+3].[La+3]
Safety and Handling

Safety and Handling

RTECSNumber : OE5400000

TSCA : TSCA

SDS Documents