missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum(III) oxide, REacton™, 99.999% (REO), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1126530

 View more versions of this product

Catalog No. AA1126530



Specifications

Specifications

Lanthanum(III) oxide
99.999% (REO)
La2O3
14,5363
Powder
Reacton™
Odorless
4200°C
250g
1312-81-8
(REO)
MFCD00011071
325.82
99.999%
6.51g/mL
Hygroscopic
2307°C

Chemical Identifiers

1312-81-8
La2O3
MFCD00011071
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : OE5330000

TSCA : TSCA

Documents