missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum(III) oxide, 99.99% (REO), Ca 10ppm max., AAS Grade, Alfa Aesar™

Manufacturer:  Alfa Aesar 11266A1

 View more versions of this product

Catalog No. AA11266A1



Specifications

Specifications

Lanthanum(III) oxide
1312-81-8
La2O3
White powder/lump
Powder
Odorless
1kg
For AAS analysis
99.99% (REO)
MFCD00011071
325.82
AAS
White

Chemical Identifiers

1312-81-8
La2O3
MFCD00011071
Safety and Handling

Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 215-200-5

Documents