missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum(III) Oxide 91%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 413022500

 View more versions of this product

Catalog No. AC413022500


 

Specifications Description & Specifications

Specifications

≥89%
Powder
White to slightly gray
MFCD00011071
91%
Lanthanum(III) oxide
La2O3
325.82
250g
15, 5414
La2O3
1312-81-8
91%
Glass bottle

Chemical Identifiers

1312-81-8
La2O3
325.82
Lanthanum(III) oxide, 91%
91%
La2O3
MFCD00011071
≥89%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 215-200-5

RTECSNumber : OE5330000

TSCA : TSCA