Please login to your online account to display your discounted pricing

Lanthanum(III) Oxide 91%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  413020050

 View more versions of this product

Catalog No. AC413020050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥89%
Formula Weight 325.82
Physical Form Powder
Quantity 5g
Color White to slightly gray
TSCA TSCA
RTECS Number OE5330000
EINECS Number 215-200-5
Merck Index 15, 5414
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Health Hazard 1 HYGROSCOPIC
Chemical Name or Material Lanthanum(III) oxide
Linear Formula La2O3
CAS 1312-81-8
Assay Percent Range 91%
MDL Number MFCD00011071
Molecular Formula La2O3
Name Note 91%

Specifications

Chemical Name or Material Lanthanum(III) oxide
Name Note 91%
CAS 1312-81-8
Assay ≥89%
Assay Percent Range 91%
Molecular Formula La2O3
Linear Formula La2O3
Formula Weight 325.82
MDL Number MFCD00011071
Accessories

HYGROSCOPIC

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.

SDS & Safety Documents