Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum(III) oxalate decahydrate, REacton™, 99.99% (REO)

Chemical Identifiers

Chemical Name or Material Lanthanum(III) oxalate decahydrate
CAS 537-03-1
Purity Grade Reacton™
Linear Formula La2(C2O4)3·10H2O
Formula Weight 722.02 (541.87 Anhydrous)
MDL Number MFCD00150540
UN Number UN3288
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1319618

  SDS
alfa aesar™
1319618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1319630

  SDS
alfa aesar™
1319630
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 722.02 (541.87 Anhydrous)
Chemical Name or Material Lanthanum(III) oxalate decahydrate
TSCA TSCA
Purity Grade Reacton™
Physical Form Powder
Assay 99.99%
Sensitivity Hygroscopic
Assay Percent Notes (REO)
Linear Formula La2(C2O4)3·10H2O
MDL Number MFCD00150540
CAS 537-03-1
UN Number UN3288
Color White
Quantity 50g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
Safety and Handling

Warning

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.