missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum(III) nitrate hydrate, 99%, water ca 22-25%

$71.57 - $252.91

Chemical Identifiers

CAS 100587-94-8
Molecular Formula H2LaN3O10
Molecular Weight (g/mol) 342.932
MDL Number MFCD00011067
InChI Key HVMFKXBHFRRAAD-UHFFFAOYSA-N
Synonym acmc-1c39v
PubChem CID 45051674
IUPAC Name lanthanum(3+);trinitrate;hydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[La+3]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1489022 View Documents Alfa Aesar™
A1489022
100g Each for $71.57
 
AAA1489036 View Documents Alfa Aesar™
A1489036
500g Each for $252.91
 
Description

Description

Applications
Lanthanum(III) nitrate hydrate serves as a raw material for the electrochemical synthesis of LaMnO4, which is a thin film coating on stainless steel substrate. It is also used in preparation of La2O2S. Further, it is used in phosphors, fluorescent display tubes and heat transfer measurement.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong reducing agents and strong acids.
Specifications

Chemical Identifiers

100587-94-8
342.932
HVMFKXBHFRRAAD-UHFFFAOYSA-N
45051674
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[La+3]
H2LaN3O10
MFCD00011067
acmc-1c39v
lanthanum(3+);trinitrate;hydrate

Specifications

100587-94-8
La(NO3)3·xH2O
UN1477
acmc-1c39v
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[La+3]
342.932
324.92 (Anhydrous)
40°C
Lanthanum(III) nitrate hydrate
H2LaN3O10
MFCD00011067
14,5363
HVMFKXBHFRRAAD-UHFFFAOYSA-N
lanthanum(3+);trinitrate;hydrate
45051674
99%
100g
water ≈22 to 25%
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

RTECSNumber : OE5075000

TSCA : TSCA