missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum(III) nitrate hydrate, 99%, water ca 22-25%

Manufacturer:  Alfa Aesar™ A1489036

 View more versions of this product

Catalog No. AAA1489036



Description

Description

Applications
Lanthanum(III) nitrate hydrate serves as a raw material for the electrochemical synthesis of LaMnO4, which is a thin film coating on stainless steel substrate. It is also used in preparation of La2O2S. Further, it is used in phosphors, fluorescent display tubes and heat transfer measurement.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong reducing agents and strong acids.
Specifications

Specifications

Lanthanum(III) nitrate hydrate
water ≈22 to 25%
La(NO3)3·xH2O
UN1477
lanthanum iii nitrate hydrate, lanthanum nitrate hydrate, nitric acid,lanthanum 3+ salt, hydrate 9ci, acmc-1c39v, la.3no3.h2o, lanthanum nitrate-water 1/3/1, lanthanum 3+ hydrate trinitrate, lanthanum 3+ ion hydrate trinitrate, lanthanum nitrate, releasing agent solution, specpure, lanthanum iii nitrate hydrate trace metals basis
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[La+3]
342.932
324.92 (Anhydrous)
40°C
100587-94-8
H2LaN3O10
MFCD00011067
14,5363
HVMFKXBHFRRAAD-UHFFFAOYSA-N
lanthanum(3+);trinitrate;hydrate
45051674
99%
500g

Chemical Identifiers

100587-94-8
342.932
HVMFKXBHFRRAAD-UHFFFAOYSA-N
45051674
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[La+3]
H2LaN3O10
MFCD00011067
lanthanum iii nitrate hydrate, lanthanum nitrate hydrate, nitric acid,lanthanum 3+ salt, hydrate 9ci, acmc-1c39v, la.3no3.h2o, lanthanum nitrate-water 1/3/1, lanthanum 3+ hydrate trinitrate, lanthanum 3+ ion hydrate trinitrate, lanthanum nitrate, releasing agent solution, specpure, lanthanum iii nitrate hydrate trace metals basis
lanthanum(3+);trinitrate;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

RTECSNumber : OE5075000

TSCA : TSCA

SDS Documents