missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum(III) nitrate hexahydrate, Reacton™, 99.999% (REO), Alfa Aesar™

$93.92 - $811.15

Chemical Identifiers

CAS 10277-43-7
Molecular Formula H12LaN3O15
Molecular Weight (g/mol) 433.007
MDL Number MFCD00149751
InChI Key GJKFIJKSBFYMQK-UHFFFAOYSA-N
Synonym kj5g6nyf2w
PubChem CID 16211478
IUPAC Name lanthanum(3+);trinitrate;hexahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[La+3]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4434614 View Documents Alfa Aesar
4434614
25g Each for $93.92
 
AA4434622 View Documents Alfa Aesar
4434622
100g Each for $214.35
 
AA4434636 View Documents Alfa Aesar
4434636
500g Each for $811.15
 
Specifications

Chemical Identifiers

10277-43-7
433.007
GJKFIJKSBFYMQK-UHFFFAOYSA-N
16211478
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[La+3]
H12LaN3O15
MFCD00149751
kj5g6nyf2w
lanthanum(3+);trinitrate;hexahydrate

Specifications

10277-43-7
H12LaN3O15
White crystalline
GJKFIJKSBFYMQK-UHFFFAOYSA-N
lanthanum(3+);trinitrate;hexahydrate
16211478
Crystalline
White
Lanthanum(III) nitrate hexahydrate
99.999% (REO)
MFCD00149751
kj5g6nyf2w
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[La+3]
433.007
433.01 (324.92 Anhydrous)
Odorless
25g
Safety and Handling

Safety and Handling

H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 5.1, Packing Group: III

EINECSNumber : 233-238-0