missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum(III) nitrate hexahydrate, Reacton™, 99.999% (REO), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4434622

 View more versions of this product

Catalog No. AA4434622



Specifications

Specifications

Lanthanum(III) nitrate hexahydrate
99.999% (REO)
MFCD00149751
kj5g6nyf2w
GJKFIJKSBFYMQK-UHFFFAOYSA-N
lanthanum(3+);trinitrate;hexahydrate
16211478
Crystalline
White
100g
10277-43-7
H12LaN3O15
White crystalline
Soluble in water,alcohol and acids.
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[La+3]
433.007
433.01 (324.92 Anhydrous)
Odorless
40°C

Chemical Identifiers

10277-43-7
433.007
GJKFIJKSBFYMQK-UHFFFAOYSA-N
16211478
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[La+3]
H12LaN3O15
MFCD00149751
kj5g6nyf2w
lanthanum(3+);trinitrate;hexahydrate
Safety and Handling

Safety and Handling

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

DOTInformation : DOT Class: 5.1; Packaging Group: II

EINECSNumber : 233-238-0

RTECSNumber : OE5075000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents