Learn More
Lanthanum(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 011267A1
Description
Lanthanum(III) nitrate is utilized as a starting material for the electrochemical synthesis of a LaMnO3 thin film coating on stainless steel substrates. It is used as a catalyst for the chemoselective preparation of cyclic and acyclic dithioacetals. It is used to prepare lanthanum aluminate (LAO) thin films, LaF3 nanocrystals, and bis(indolyl) methanes from indoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lanthanum(III) nitrate hexahydrate | |
10277-43-7 | |
Crystalline Aggregates | |
99.99% (REO) | |
MFCD00149751 | |
Soluble in water,alcohol and acids. | |
O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
433.01 | |
433.01 (324.92 Anhydrous) | |
White crystalline |
40°C | |
White | |
1 kg | |
H12LaN3O15 | |
lanthanum iii nitrate hexahydrate, lanthanum nitrate hexahydrate, unii-kj5g6nyf2w, kj5g6nyf2w, lanthanum iii nitrate, hexahydrate 1:3:6, lanthanum nitrate, 6-hydrate, ksc909a4f, lanthanum nitrate hexahydrate,, lanthanum 3+ hexahydrate trinitronate | |
GJKFIJKSBFYMQK-UHFFFAOYSA-N | |
lanthanum(3+);trinitrate;hexahydrate | |
16211478 | |
Odorless |
Chemical Identifiers
10277-43-7 | |
433.01 | |
GJKFIJKSBFYMQK-UHFFFAOYSA-N | |
16211478 |
H12LaN3O15 | |
MFCD00149751 | |
lanthanum iii nitrate hexahydrate, lanthanum nitrate hexahydrate, unii-kj5g6nyf2w, kj5g6nyf2w, lanthanum iii nitrate, hexahydrate 1:3:6, lanthanum nitrate, 6-hydrate, ksc909a4f, lanthanum nitrate hexahydrate,, lanthanum 3+ hexahydrate trinitronate | |
O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Safety and Handling
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335
DOTInformation : DOT Class: 5.1; Packaging Group: II
EINECSNumber : 233-238-0
RTECSNumber : OE5075000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only