missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01126730

 View more versions of this product

Catalog No. AA1126730



Description

Description

Lanthanum(III) nitrate is utilized as a starting material for the electrochemical synthesis of a LaMnO3 thin film coating on stainless steel substrates. It is used as a catalyst for the chemoselective preparation of cyclic and acyclic dithioacetals. It is used to prepare lanthanum aluminate (LAO) thin films, LaF3 nanocrystals, and bis(indolyl) methanes from indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lanthanum(III) nitrate hexahydrate
10277-43-7
Crystalline Aggregates
99.99% (REO)
MFCD00149751
Soluble in water,alcohol and acids.
O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
433.01
433.01 (324.92 Anhydrous)
White crystalline
40°C
White
250 g
H12LaN3O15
lanthanum iii nitrate hexahydrate, lanthanum nitrate hexahydrate, unii-kj5g6nyf2w, kj5g6nyf2w, lanthanum iii nitrate, hexahydrate 1:3:6, lanthanum nitrate, 6-hydrate, ksc909a4f, lanthanum nitrate hexahydrate,, lanthanum 3+ hexahydrate trinitronate
GJKFIJKSBFYMQK-UHFFFAOYSA-N
lanthanum(3+);trinitrate;hexahydrate
16211478
Odorless

Chemical Identifiers

10277-43-7
433.01
GJKFIJKSBFYMQK-UHFFFAOYSA-N
16211478
H12LaN3O15
MFCD00149751
lanthanum iii nitrate hexahydrate, lanthanum nitrate hexahydrate, unii-kj5g6nyf2w, kj5g6nyf2w, lanthanum iii nitrate, hexahydrate 1:3:6, lanthanum nitrate, 6-hydrate, ksc909a4f, lanthanum nitrate hexahydrate,, lanthanum 3+ hexahydrate trinitronate
O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Safety and Handling

Safety and Handling

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

DOTInformation : DOT Class: 5.1; Packaging Group: II

EINECSNumber : 233-238-0

RTECSNumber : OE5075000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only