missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum(III) nitrate hexahydrate, 99.9% (REO), Alfa Aesar™

Manufacturer:  Alfa Aesar 1291536

 View more versions of this product

Catalog No. AA1291536



Specifications

Specifications

Lanthanum(III) nitrate hexahydrate
99.9% (REO)
H12LaN3O15
UN1477
kj5g6nyf2w
GJKFIJKSBFYMQK-UHFFFAOYSA-N
lanthanum(3+);trinitrate;hexahydrate
16211478
Crystalline
Odorless
500g
10277-43-7
(REO)
MFCD00149751
14,5363
Soluble in water and alcohol
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[La+3]
433.007
433.01 (324.92 Anhydrous)
99.9%
40°C

Chemical Identifiers

10277-43-7
433.007
GJKFIJKSBFYMQK-UHFFFAOYSA-N
16211478
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[La+3]
H12LaN3O15
MFCD00149751
kj5g6nyf2w
lanthanum(3+);trinitrate;hexahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

RTECSNumber : OE5075000

TSCA : TSCA

Documents